3-Fluoro-4-formylpyridine 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3-Fluoro-4-pyridinecarbaldehyde, 3-Fluoroisonicotinaldehyde, 3-Fluoro-4-pyridinecarboxaldehyde
442170010 442170050
ACRO442170010EA 1275 SEK
ACRO442170010 ACRO442170050
3-Fluoro-4-formylpyridine 97%
3-Fluoro-4-formylpyridine
Formel: C₆H₄FNO
MW: 125.1 g/mol
Kokpunkt: 70 °C (25Torr)
Densitet: 1.255 g/cm³
Flampunkt: 67 °C
Storage Temperature: Omgivande
MDL Number: MFCD06410686
CAS nummer: 40273-47-0
Order Now
Specifikation testresultat
Appearance Clear colorless to orange to brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %