2-Chloro-4-fluoropyridine 97%
Leverantör: Thermo Fisher Scientific
Fara
442150010 442150050
ACRO442150010EA 1925 SEK
ACRO442150010 ACRO442150050
2-Chloro-4-fluoropyridine 97%
2-Chloro-4-fluoropyridine
Formel: C₅H₃ClFN
Kokpunkt: 140 °C (760 mmHg)
Densitet: 1,456 g/cm³
Flampunkt: 54 °C
Storage Temperature: Omgivande
MDL Number: MFCD04039345
CAS nummer: 34941-91-8
UN: 2929
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %