1-Aminohydantoin hydrochloride 98%
Leverantör: Thermo Fisher Scientific
Synonyms: 1-Aminohydantoin Hydrochloride, 1-Aminohydantoin HCl, 1-Amino-2,4-imidazolidinedione hydrochloride
442050050 442050250
ACRO442050050EA 830 SEK
ACRO442050050 ACRO442050250
1-Aminohydantoin hydrochloride 98%
1-Aminohydantoin hydrochloride
Formel: C₃H₅N₃O₂
MW: 151.55 g/mol
Smältpunkt: 201…205 °C
Storage Temperature: Omgivande
MDL Number: MFCD02181047
CAS nummer: 2827-56-7
Order Now
Specifikation testresultat
Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Titration Argentometric 97.5 to 102.5 %
HPLC ≥97.5 %