tert-Butyl-3-oxoazetidine-1-carboxylate 97%
Leverantör: Thermo Scientific
Varning
Synonyms: N-Boc-3-azetidinone, Tert-butyl 3-oxoazetidine-1-carboxylate, tert-Butyl 3-oxoazetidine-1-carboxylate, 1-BOC-3-Azetidinone
442040010 442040050
ACRO442040010EA 1500 SEK
ACRO442040010 ACRO442040050
tert-Butyl-3-oxoazetidine-1-carboxylate 97%
tert-Butyl-3-oxoazetidine-1-carboxylate
Formel: C₈H₁₃NO₃
MW: 171,2 g/mol
Smältpunkt: 50…52 °C
Storage Temperature: Omgivande
MDL Number: MFCD01861741
CAS nummer: 398489-26-4
UN: 3335
ADR: 9,III
Order Now
Specifikation testresultat
Appearance White to light brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 48 to 54 °C
GC ≥96.0 %