N-Boc-cis-4-hydroxy-L-proline 97%
Leverantör: Thermo Scientific
Varning
Synonyms: N-BOC-cis-4-Hydroxy-L-proline, cis-N-(tert-Butoxycarbonyl)-4-hydroxy-L-proline, Boc-cis-Hyp-OH, cis-N-Boc-4-hydroxy-L-proline, Boc-Cis-Hyp-OH, (2S,4S)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid
441880010 441880050
ACRO441880010EA 1525 SEK
ACRO441880010 ACRO441880050
N-Boc-cis-4-hydroxy-L-proline 97%
N-Boc-cis-4-hydroxy-L-proline
Formel: C₁₀H₁₇NO₅
MW: 231,248 g/mol
Smältpunkt: 137…143 °C
Storage Temperature: Omgivande
MDL Number: MFCD02094406
CAS nummer: 87691-27-8
UN: 3077
ADR: 9,III
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
Specific optical rotation -47° to -53° (20°C, 589 nm) (c=0.67 in MeOH)
Titration with NaOH 96 to 104 %