4-Cyanopiperidine 97%
Leverantör: Thermo Fisher Scientific
Fara
441820050 441820250
ACRO441820050EA 970 SEK
ACRO441820050 ACRO441820250
4-Cyanopiperidine 97%
4-Cyanopiperidine
Formel: C₆H₁₀N₂
Kokpunkt: 100…105 °C (12 mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD05022468
CAS nummer: 4395-98-6
UN: 3276
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %