1-Methyl-4-piperidinemethanol 97%
Leverantör: Thermo Scientific
Varning
440480010 440480050
ACRO440480010EA 740 SEK
ACRO440480010 ACRO440480050
1-Methyl-4-piperidinemethanol 97%
1-Methyl-4-piperidinemethanol
Formel: C₇H₁₅NO
Smältpunkt: 29…31 °C
Densitet: 0,98 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD03411903
CAS nummer: 20691-89-8
Order Now
Specifikation testresultat
Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
Titration with HCl ≥96.0 to ≤104.0 %