1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester 97%
Leverantör: Thermo Fisher Scientific
Varning
439730010 439730050
ACRO439730010EA 1175 SEK
ACRO439730010 ACRO439730050
1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester 97%
Formel: C₁₃H₂₃BN₂O₂
Kokpunkt: 248…249 °C (760 mmHg)
Densitet: 1,003 g/cm³
Flampunkt: 48 °C
Storage Temperature: Omgivande
MDL Number: MFCD05663857
CAS nummer: 827614-66-4
UN: 1993
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %