(R)-1-Methyl-3-pyrrolidinol 99%
Leverantör: Thermo Scientific
Varning
Synonyms: (R)-3-Hydroxy-1-methylpyrrolidine, (R)-(-)-1-Methyl-3-pyrrolidinol, (R)-3-Hydroxy-1-methyl-pyrrolidine
439440010 439440050
ACRO439440010EA 1200 SEK
ACRO439440010 ACRO439440050
(R)-1-Methyl-3-pyrrolidinol 99%
(R)-1-Methyl-3-pyrrolidinol
Formel: C₅H₁₁NO
MW: 101,15 g/mol
Kokpunkt: 50…52 °C (1 mmHg)
Densitet: 0,921 g/cm³
Flampunkt: 70 °C
Storage Temperature: Omgivande
MDL Number: MFCD03788747
CAS nummer: 104641-60-3
Order Now
Specifikation testresultat
Appearance Colorless to yellow to brown Liquid (May darken on storage)
Infrared spectrum Conforms
Specific optical rotation -6° to -8° (20°C, 589 nm) (c=1 in chloroform)
Titration with HClO4 98.5 to 101.5 %