4-Bromophenethyl bromide 96%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-Bromo-4-(2-bromoethyl)benzene
439530010 439530100
ACRO439530010EA 745 SEK
ACRO439530010 ACRO439530100
4-Bromophenethyl bromide 96%
4-Bromophenethyl bromide
Formel: C₈H₈Br₂
MW: 263.96 g/mol
Kokpunkt: 97…98.5 °C (1 mmHg)
Densitet: 1.73 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00045052
CAS nummer: 1746-28-7
UN: 3082
ADR: 9 III
Order Now
Specifikation testresultat
Appearance Colorless to yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥95.0 %