Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0) 96%
Leverantör: Thermo Fisher Scientific
Varning
439150010 439150050
ACRO439150010EA 1100 SEK
ACRO439150010 ACRO439150050
Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0) 96%
Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0)
Formel: C₄₂H₄₄O₁₀Pd
Smältpunkt: 170…178 °C
Storage Temperature: Kylskåp
MDL Number: MFCD07369799
CAS nummer: 811862-77-8
Order Now
Specifikation testresultat
Appearance Red-brown Powder
Infrared spectrum Conforms
NMR ≥95.0 %