5-Iodo-2-methoxypyridine 97%
Leverantör: Thermo Scientific
Varning
438930010 438930050
ACRO438930010EA 2275 SEK
ACRO438930010 ACRO438930050
5-Iodo-2-methoxypyridine 97%
5-Iodo-2-methoxypyridine
Formel: C₆H₆INO
MW: 235,03 g/mol
Kokpunkt: 106…108 °C (30Torr)
Storage Temperature: Omgivande
MDL Number: MFCD07781180
CAS nummer: 13472-61-2
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %