1,4-Diazabicyclo[3.2.2]nonane 95%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: N-phenyl-1,4-diazabicyclo[3.2.2]nonane-4-carboxamide, 1,4-diaza-bicyclo[3.2.2]nonane, 1,4-Diazobicylco[3.2.2]nonane
438900010 438900050
ACRO438900010EA 4175 SEK
ACRO438900010 ACRO438900050
1,4-Diazabicyclo[3.2.2]nonane 95%
1,4-Diazabicyclo[3.2.2]nonane
Formel: C₇H₁₄N₂
MW: 126.2 g/mol
Kokpunkt: 77…79 °C (6Torr)
Smältpunkt: 58…79 °C
Storage Temperature: Omgivande
MDL Number: MFCD14581694
CAS nummer: 283-38-5
UN: 3259
ADR: 8 III
Order Now
Specifikation testresultat
Appearance Colorless to brown Liquid or waxy solid
Infrared spectrum Conforms
GC ≥94.0 %