(1,3-Dioxolan-2-ylmethyl)magnesium bromide 0.5 M i THF, AcroSeal®
Leverantör: Thermo Scientific
Fara
Synonyms: Magnesium 2-methanidyl-1,3-dioxolane Bromide, magnesium(2+) ion 1,3-dioxolan-2-ylmethanide bromide
438861000 438868000
438861000.EA 2000 SEK
438861000. 438868000.
(1,3-Dioxolan-2-ylmethyl)magnesium bromide 0.5 M i THF, AcroSeal®
(1,3-Dioxolan-2-ylmethyl)magnesium bromide
Formel: C₄H₇BrMgO₂
MW: 191,31 g/mol
Kokpunkt: 65…67 °C (760 mmHg)
Densitet: 0,94 g/cm³
Flampunkt: –17 °C
Storage Temperature: Omgivande
MDL Number: MFCD01321271
CAS nummer: 180675-22-3
UN: 2924
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Light yellow to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 0.40 to 0.60 M