(3-Bromopropoxy)-tert-butyldimethylsilane 97% stabilised
Leverantör: Thermo Scientific
Varning
Synonyms: (3-Bromopropoxy)(tert-butyl)dimethylsilane
Stabilization: Stabilised with sodium carbonate (Na₂CO₃)
438690050 438690250
ACRO438690050EA 2000 SEK
ACRO438690050 ACRO438690250
(3-Bromopropoxy)-tert-butyldimethylsilane 97% stabilised
(3-Bromopropoxy)-tert-butyldimethylsilane
Formel: C₉H₂₁BrOSi
MW: 253,26 g/mol
Kokpunkt: 182 °C (760 mmHg)
Densitet: 1,093 g/cm³
Flampunkt: 85 °C
Storage Temperature: Omgivande
MDL Number: MFCD00216589
CAS nummer: 89031-84-5
Order Now
Specifikation testresultat
Appearance Colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %