3-Ethynylthiophene 96%
Leverantör: Thermo Scientific
Fara
438580010 438580050
ACRO438580010EA 1275 SEK
ACRO438580010 ACRO438580050
3-Ethynylthiophene 96%
3-Ethynylthiophene
Formel: C₆H₄S
MW: 108,16 g/mol
Kokpunkt: 152…154 °C (760 mmHg)
Densitet: 1,098 g/cm³
Flampunkt: 46 °C
Storage Temperature: Kylskåp
MDL Number: MFCD04039973
CAS nummer: 67237-53-0
UN: 1993
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Colorless to yellow to orange Liquid (May darken on storage)
Infrared spectrum Conforms
GC ≥95.0 %