N-Boc-pyrrole 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1H-pyrrole-1-carboxylic acid, 1,1-dimethylethyl ester, N-(tert-butyloxycarbonyl)pyrrole, tert-Butyl 1-pyrrolecarboxylate
438510250 438511000
ACRO438510250EA 1400 SEK
ACRO438510250 ACRO438511000
N-Boc-pyrrole 98%
N-Boc-pyrrole
Formel: C₉H₁₃NO₂
Kokpunkt: 97 °C (2,3Torr)
Densitet: 1 g/cm³
Flampunkt: 75 °C
Storage Temperature: Omgivande
MDL Number: MFCD00209559
CAS nummer: 5176-27-2
Order Now
Specifikation testresultat
Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
GC ≥97.5 %