4,4,5,5,5-Pentafluoropentan-1-ol 95%
Leverantör: Thermo Scientific
Varning
Synonyms: 4,4,5,5,5-Pentafluoropentanol
438430050 438430250
ACRO438430050EA 1600 SEK
ACRO438430050 ACRO438430250
4,4,5,5,5-Pentafluoropentan-1-ol 95%
4,4,5,5,5-Pentafluoropentan-1-ol
Formel: C₅H₇F₅O
MW: 178,1 g/mol
Kokpunkt: 134…135 °C
Flampunkt: 63 °C
Storage Temperature: Omgivande
MDL Number: MFCD00153224
CAS nummer: 148043-73-6
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥94.0 %