Methyl benzenesulphinate 98%
Leverantör: Thermo Scientific
Varning
438610050 438610250
ACRO438610050EA 1925 SEK
ACRO438610050 ACRO438610250
Methyl benzenesulphinate 98%
Methyl benzenesulphinate
Formel: C₇H₈O₂S
MW: 156,21 g/mol
Kokpunkt: 110…112 °C (16Torr)
Densitet: 1,194 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00674051
CAS nummer: 670-98-4
Order Now
Specifikation testresultat
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %