1-Boc-piperidine 97%
Leverantör: Thermo Scientific
Varning
Synonyms: tert-Butyl Piperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)piperidine, Piperidine-1-carboxylic acid tert-Butyl ester, tert-butyl piperidine-1-carboxylate, 1-Boc-piperidine
438080050 438080250
ACRO438080050EA 800 SEK
ACRO438080050 ACRO438080250
1-Boc-piperidine 97%
1-Boc-piperidine
Formel: C₁₀H₁₉NO₂
MW: 185,27 g/mol
Kokpunkt: 58 °C (,3mbar)
Flampunkt: 86 °C
Storage Temperature: Kylskåp
MDL Number: MFCD02093935
CAS nummer: 75844-69-8
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %