2-Aminopyridine-4-methanol 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Amino-4-pyridinylmethanol, (2-Aminopyridin-4-yl)-methanol, (2-Aminopyridin-4-yl)methanol, (2-Aminopyrid-4-yl)methanol
437370010 437370050
ACRO437370010EA 1450 SEK
ACRO437370010 ACRO437370050
2-Aminopyridine-4-methanol 97%
2-Aminopyridine-4-methanol
Formel: C₆H₈N₂O
MW: 124,143 g/mol
Smältpunkt: 75,5…84 °C
Storage Temperature: Omgivande
MDL Number: MFCD03791261
CAS nummer: 105250-17-7
Order Now
Specifikation testresultat
Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %