DL-Propargylglycine 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2-Aminopent-4-ynoic acid, PAG, 2-Amino-4-pentynoic acid, H-DL-Pra-OH
437320010 437320050
ACRO437320010EA 2275 SEK
ACRO437320010 ACRO437320050
DL-Propargylglycine 98%
DL-Propargylglycine
Formel: C₅H₇NO₂
MW: 113.116 g/mol
Smältpunkt: 235 °C
Storage Temperature: Frys
MDL Number: MFCD00056728
CAS nummer: 64165-64-6
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %