(S)-1-(Diphenylphosphino)-2-[(S)-4-isopropyloxazolin-2-yl]ferrocene 97%
Leverantör: Thermo Scientific
Synonyms: (S,S)-ip-FOXAP, (S,S)-[2-(4-Isopropyl-2-oxazolinyl)ferrocenyl]diphenylphosphine, (S,S)-[2-(4'-Isopropyloxazolin-2'yl)ferrocenyl]diphenylphosphine
437281000 437282500
ACRO437281000EA 1600 SEK
ACRO437281000 ACRO437282500
(S)-1-(Diphenylphosphino)-2-[(S)-4-isopropyloxazolin-2-yl]ferrocene 97%
(S)-1-(Diphenylphosphino)-2-[(S)-4-isopropyloxazolin-2-yl]ferrocene
Formel: C₂₈H₂₈FeNOP
MW: 481,36 g/mol
Smältpunkt: 162…165 °C
Storage Temperature: Kylskåp
MDL Number: MFCD15144869
CAS nummer: 163169-29-7
UN: 3335
ADR: 9,III
Order Now
Specifikation testresultat
Appearance Yellow to brown Crystalline powder or crystals
Infrared spectrum Conforms
HPLC ≥96.0 %