Natriumnitrat 99.999%
Leverantör: Thermo Scientific
Varning
437160100 437160500 437165000
ACRO437160100EA 1175 SEK
ACRO437160100 ACRO437160500 ACRO437165000
Natriumnitrat 99.999%
Sodium nitrate
Formel: NaNO₃
MW: 84,99 g/mol
Kokpunkt: 380 °C (760 mmHg)
Smältpunkt: 306 °C
Densitet: 2,26 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011119
CAS nummer: 7631-99-4
UN: 1498
ADR: 5.1,III
Order Now
Specifikation testresultat
Appearance White Powder or crystals
Total trace metal impurities ≤10 ppm
Sodium (Na) Confirmed