Tellur 99.999%, pulver
Leverantör: Thermo Scientific
Varning
437010250 437011000
ACRO437010250EA 1525 SEK
ACRO437010250 ACRO437011000
Tellur 99.999%, pulver
Tellurium
Formel: Te
MW: 127,6 g/mol
Kokpunkt: 990 °C (760 mmHg)
Smältpunkt: 450 °C
Densitet: 8,83 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134062
CAS nummer: 13494-80-9
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Grey to black Powder
Total trace metal impurities ≤10 ppm