Indium(III) acetate, vattenfri 99.99%
Leverantör: Thermo Scientific
Varning
436980100 436980500
ACRO436980100EA 1625 SEK
ACRO436980100 ACRO436980500
Indium(III) acetate, vattenfri 99.99%
Indium(III) acetate
Formel: C₂H₄InO₂
MW: 174,87 g/mol
Smältpunkt: 270 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00054347
CAS nummer: 25114-58-3
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Titration Complexometric 35.3 to 43.3 % (In)
Total trace metal impurities ≤100 ppm