1-Benzyl-3-pyrroline 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 1-Benzyl-2,5-dihydropyrrole
436770010 436770050
ACRO436770010EA 560 SEK
ACRO436770010 ACRO436770050
1-Benzyl-3-pyrroline 97%
1-Benzyl-3-pyrroline
Formel: C₁₁H₁₃N
Kokpunkt: 92 °C (1hPa)
Densitet: 0,837 g/cm³
Flampunkt: 92 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012324
CAS nummer: 6913-92-4
UN: 3267
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %