3-Bromo-2-methylpyridine 97%
Leverantör: Thermo Fisher Scientific
Fara
436860010 436860050
ACRO436860010EA 1325 SEK
ACRO436860010 ACRO436860050
3-Bromo-2-methylpyridine 97%
3-Bromo-2-methylpyridine
Formel: C₆H₆BrN
Kokpunkt: 65…67 °C (10Torr)
Densitet: 1,495 g/cm³
Flampunkt: 79 °C
Storage Temperature: Omgivande
MDL Number: MFCD00191224
CAS nummer: 38749-79-0
Order Now
Specifikation testresultat
Appearance Clear colorless to brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %