(S)-(-)-3-Aminotetrahydrofuran-p-toluenesulphonate salt 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: (S)-(-)-Tetrahydro-3-furylamine p-Toluenesulphonate salt, (S)-Tetrahydrofuran-3-amine-4-methylbenzenesulphonate, 3-Furanamine, tetrahydro-, (3S)-, 4-methylbenzenesulphonate (1:1)
435830010 435830050
ACRO435830010EA 935 SEK
ACRO435830010 ACRO435830050
(S)-(-)-3-Aminotetrahydrofuran-p-toluenesulphonate salt 97%
(S)-(-)-3-Aminotetrahydrofuran-p-toluenesulphonate salt
Formel: C₁₁H₁₇NO₄S
MW: 259.32 g/mol
Smältpunkt: 137.9…142.8 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00066238
CAS nummer: 104530-79-2
UN: 2811
ADR: 6.1 III
Order Now
Specifikation testresultat
Appearance White to off-white to light brown Crystals or solid
Infrared spectrum Conforms
Elemental Analysis 5.2 to 5.6 % N
Specific optical rotation -3° to -5° (20°C, 589 nm) (c=1, methanol)