(S)-(-)-1-Boc-3-aminopyrrolidine 95%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: (S)-1-Boc-3-Aminopyrrolidine, (S)-(-)-1-BOC-3-aminopyrrolidine, tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate
435450010 435450050
ACRO435450010EA 1475 SEK
ACRO435450010 ACRO435450050
(S)-(-)-1-Boc-3-aminopyrrolidine 95%
(S)-(-)-1-Boc-3-aminopyrrolidine
Formel: C₉H₁₈N₂O₂
MW: 186.25 g/mol
Kokpunkt: 120…125 °C (5Torr)
Flampunkt: 91 °C
Storage Temperature: Omgivande
MDL Number: MFCD03419271
CAS nummer: 147081-44-5
UN: 2810
ADR: 6.1 III
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %
Specific optical rotation -1.5° to -3.0° (20°C, 589 nm) (C=1, chloroform)