4-Fluoro-3-methylphenylboronic acid 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Methyl-4-fluorophenylboronic acid, 4-Fluoro-3-methylbenzeneboronic acid
435330010 435330050 435330250
ACRO435330010EA 320 SEK
ACRO435330010 ACRO435330050 ACRO435330250
4-Fluoro-3-methylphenylboronic acid 98%
4-Fluoro-3-methylphenylboronic acid
Formel: C₇H₈BFO₂
MW: 153,95 g/mol
Smältpunkt: 215,4…217.3 °C
Storage Temperature: Omgivande
MDL Number: MFCD01863527
CAS nummer: 139911-27-6
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
GC ≥96.0 %