4-Ethoxybenzoyl chloride 98%
Leverantör: Thermo Fisher Scientific
Fara
434970010 434970050
ACRO434970010EA 845 SEK
ACRO434970010 ACRO434970050
4-Ethoxybenzoyl chloride 98%
4-Ethoxybenzoyl chloride
Formel: C₉H₉ClO₂
Kokpunkt: 128…130 °C (6Torr)
Flampunkt: 113 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000688
CAS nummer: 16331-46-7
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Colorless to yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥97.5 %