2,4-Dichloro-5-nitropyrimidine 97%
Leverantör: Thermo Fisher Scientific
Varning
435290050 435290250
ACRO435290050EA 1475 SEK
ACRO435290050 ACRO435290250
2,4-Dichloro-5-nitropyrimidine 97%
2,4-Dichloro-5-nitropyrimidine
Formel: C₄HCl₂N₃O₂
MW: 193.98 g/mol
Kokpunkt: 135 °C (17Torr)
Smältpunkt: 29…30 °C
Densitet: 1.74
Storage Temperature: Omgivande
MDL Number: MFCD00127867
CAS nummer: 49845-33-2
Order Now
Specifikation testresultat
Appearance Clear yellow Liquid or low melting solid
Infrared spectrum Conforms
Melting point 28 to 32 °C
GC ≥96.0 %