β-Alanine-tert-butylester hydrochloride 98%
Leverantör: Thermo Scientific
Synonyms: beta-Alanine tert-Butyl ester hydrochloride, H-β-Ala-OtBu·HCl, H-beta-Ala-OtBu·HCl, H-beta-Ala-OtBu.HCl, H-β-Ala-OtBu.HCl
434650010 434650050
ACRO434650010EA 2050 SEK
ACRO434650010 ACRO434650050
β-Alanine-tert-butylester hydrochloride 98%
beta-Alanine-tert-butylester hydrochloride
Formel: C₇H₁₆ClNO₂
MW: 181,66 g/mol
Smältpunkt: 167,8…168.5 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00058258
CAS nummer: 58620-93-2
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder or solid
Infrared spectrum Conforms
Elemental Analysis 7.5 to 7.9 % N
Titration with NaOH ≥98.5 %