p-Toluenesulphinic acid sodium salt, vattenfri 97%
Leverantör: Thermo Scientific
Synonyms: Sodium 4-methylbenzenesulphinate, Sodium-p-toluenesulphinate, 4-Methylbenzenesulphinic acid sodium salt, Sodiumtoluene-4-sulphinate
434530250 434531000
ACRO434530250EA 740 SEK
ACRO434530250 ACRO434531000
p-Toluenesulphinic acid sodium salt, vattenfri 97%
p-Toluenesulphinic acid sodium salt
Formel: C₇H₇NaO₂S
MW: 178,19 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00013136
CAS nummer: 824-79-3
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤3.00 %
HPLC ≥96.0 %