1-Methylbenzimidazole 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Methyl-1H-benzimidazole
434420050 434420250
ACRO434420050EA 1050 SEK
ACRO434420050 ACRO434420250
1-Methylbenzimidazole 99%
1-Methylbenzimidazole
Formel: C₈H₈N₂
MW: 132,16 g/mol
Kokpunkt: 154 °C (16hPa)
Smältpunkt: 55…57 °C
Densitet: 1,11 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00192275
CAS nummer: 1632-83-3
Order Now
Specifikation testresultat
Appearance White to pink solid
Infrared spectrum Authentic
Melting point 55 to 60 °C
GC ≥98.5 %