4-Chlorotetrahydropyran 96%
Leverantör: Thermo Fisher Scientific
Varning
434270010 434270050
ACRO434270010EA 985 SEK
ACRO434270010 ACRO434270050
4-Chlorotetrahydropyran 96%
4-Chlorotetrahydropyran
Formel: C₅H₉ClO
MW: 120.5800 g/mol
Kokpunkt: 150 °C (760 mmHg)
Densitet: 1.12 g/cm³
Flampunkt: 46 °C
Storage Temperature: Omgivande
MDL Number: MFCD00074911
CAS nummer: 1768-64-5
UN: 1993
ADR: 3 III
Order Now
Specifikation testresultat
Appearance Colorless to dark yellow Liquid
Infrared spectrum Conforms
GC ≥95.0 %