N-Methyl-1,2-phenylenediamine 97%
Leverantör: Thermo Scientific
Varning
Synonyms: N1-Methylbenzene-1,2-diamine, N-Methylbenzene-1,2-diamine, N-Methyl-o-phenylenediamine, 2-Amino-N-methylaniline
434230050 434230250
ACRO434230050EA 1950 SEK
ACRO434230050 ACRO434230250
N-Methyl-1,2-phenylenediamine 97%
N-Methyl-1,2-phenylenediamine
Formel: C₇H₁₀N₂
MW: 122,171 g/mol
Kokpunkt: 123…124 °C (10 mmHg)
Smältpunkt: 22 °C
Densitet: 1,075 g/cm³
Flampunkt: 113 °C
Storage Temperature: Omgivande
MDL Number: MFCD00192314
CAS nummer: 4760-34-3
Order Now
Specifikation testresultat
Appearance Clear yellow to red to dark brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %