4-(tert-Butoxycarbonylamino)-1-butanol 98%
Leverantör: Thermo Scientific
Varning
Synonyms: tert-Butyl (4-hydroxybutyl)carbamate, N-Boc-4-aminobutanol, 4-(Boc-amino)-1-butanol, N-(4-Hydroxybutyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(4-Hydroxybutyl)carbamate
434210010 434210050
ACRO434210010EA 1400 SEK
ACRO434210010 ACRO434210050
4-(tert-Butoxycarbonylamino)-1-butanol 98%
4-(tert-Butoxycarbonylamino)-1-butanol
Formel: C₉H₁₉NO₃
MW: 189,26 g/mol
Smältpunkt: 37…40 °C
Densitet: 1,02 g/cm³
Storage Temperature: Kylskåp
MDL Number: MFCD01861334
CAS nummer: 75178-87-9
Order Now
Specifikation testresultat
Appearance Colorless to yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥97.5 %
HPLC ≥96.0 %