2-Bromo-5-methylthiophene 95% stabilised
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 5-Methyl-2-bromothiophene, 2-Bromo-5-methylthiophenol
Stabilization: (0.1:0.4)% Stabilised with copper and sodium hydrogen carbonate
434200010 434200050
ACRO434200010EA 605 SEK
ACRO434200010 ACRO434200050
2-Bromo-5-methylthiophene 95% stabilised
2-Bromo-5-methylthiophene
Formel: C₅H₅BrS
MW: 177.06 g/mol
Kokpunkt: 65 °C (15 mmHg)
Smältpunkt: 58
Flampunkt: 78 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00130103
CAS nummer: 765-58-2
Order Now
Specifikation testresultat
Appearance Yellow Liquid (May develop some turbidity or precipitate)
Infrared spectrum Conforms
GC ≥94.0 %
Stabilizer 0.1 % Cu and 0.4 % NaHCO3