4,7-Dimethoxy-1,10-phenanthroline 97%
Leverantör: Thermo Fisher Scientific
Fara
433780010 433782500
ACRO433780010EA 5600 SEK
ACRO433780010 ACRO433782500
4,7-Dimethoxy-1,10-phenanthroline 97%
4,7-Dimethoxy-1,10-phenanthroline
Formel: C₁₄H₁₂N₂O₂
MW: 240.2573 g/mol
Smältpunkt: 212…214 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00233883
CAS nummer: 92149-07-0
UN: 3077
ADR: 9 III
Order Now
Specifikation testresultat
Appearance White to brown Crystalline powder or solid
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥95 %