Tetrabutylammonium fluoride 1 M i THF, AcroSeal®
Leverantör: Thermo Fisher Scientific
Fara
433511000 433518000
433511000.EA 2500 SEK
433511000. 433518000.
Tetrabutylammonium fluoride 1 M i THF, AcroSeal®
Tetrabutylammonium fluoride
Formel: C₁₆H₃₆FN
MW: 261.47 g/mol
Densitet: 0.887 g/cm³
Flampunkt: –17 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00011747
CAS nummer: 429-41-4
UN: 2924
ADR: 3 II
Order Now
Specifikation testresultat
Appearance Clear yellow to amber to light green to brown Solution
Water ≤8 %
Titration with HClO4 0.90 to 1.10 M