2-Mercaptoimidazole 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Imidazole-2-thiol, 2-Mercapto-1H-imidazole
433360010 433360050
ACRO433360010EA 910 SEK
ACRO433360010 ACRO433360050
2-Mercaptoimidazole 98%
2-Mercaptoimidazole
Formel: C₃H₄N₂S
MW: 100,14 g/mol
Smältpunkt: 228…231 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005188
CAS nummer: 872-35-5
UN: 3335
ADR: 9,III
Order Now
Specifikation testresultat
Appearance White to yellow to tan Crystalline powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %