3-Chlorophenyl acetylene 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Chloro-3-ethynylbenzene, 3-Chlorophenylacetylene, (3-Chlorophenyl)acetylene
433130010 433130050
ACRO433130010EA 995 SEK
ACRO433130010 ACRO433130050
3-Chlorophenyl acetylene 97%
3-Chlorophenyl acetylene
Formel: C₈H₅Cl
MW: 136,578 g/mol
Kokpunkt: 178…179 °C (760 mmHg)
Flampunkt: 63 °C
Storage Temperature: Kylskåp
MDL Number: MFCD03839984
CAS nummer: 766-83-6
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %