1H-Tetrazole 3 - 4% (w/w) i acetonitril, AcroSeal®
Leverantör: Thermo Fisher Scientific
Fara
432990010 432991000
432990010.EA 14250 SEK
432990010. 432991000.
1H-Tetrazole 3 - 4% (w/w) i acetonitril, AcroSeal®
1H-Tetrazole
Formel: CH₂N₄
Kokpunkt: 77 °C
Densitet: 0,8 g/cm³
Flampunkt: 5 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005247
CAS nummer: 288-94-8
UN: 1993
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Solution (May develop some turbidity or precipitate)
Water ≤0.3 %
Strength 3.0 to 4.0 % (wt)
Titration with HClO4 31.0 to 35.0 %