Pyrazolo[1,5-a]pyrimidine-5-carboxylic acid 97%
Leverantör: Thermo Scientific
Varning
432720010 432720050
ACRO432720010EA 3800 SEK
ACRO432720010 ACRO432720050
Pyrazolo[1,5-a]pyrimidine-5-carboxylic acid 97%
Pyrazolo[1,5-a]pyrimidine-5-carboxylic acid
Formel: C₇H₅N₃O₂
Smältpunkt: 222…225 °C
Storage Temperature: Omgivande
MDL Number: MFCD09414725
CAS nummer: 1086375-50-9
Order Now
Specifikation testresultat
Appearance Yellow Powder
Infrared spectrum Conforms
HPLC ≥96.0 %