4,4,4-Trifluorobutylamine 97%
Leverantör: Thermo Scientific
Fara
Synonyms: 1-Butanamine,4,4,4-trifluoro-, 4,4,4-trifluorobutan-1-amine, 1-Amino-4,4,4-trifluorobutane
432590010 432590050
ACRO432590010EA 1750 SEK
ACRO432590010 ACRO432590050
4,4,4-Trifluorobutylamine 97%
4,4,4-Trifluorobutylamine
Formel: C₄H₈F₃N
MW: 127,11 g/mol
Kokpunkt: 90…94 °C (760 mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD06213030
CAS nummer: 819-46-5
UN: 2734
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %
Titration RCOCl ≥96.0 %