trans-β-Styrylboronic acid pinacol ester 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: E-Phenylethenylboronic acid, pinacol ester, Trans-2-Phenylvinylboronic acid pinacol ester, trans-2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)styrene
432030010 432030050
ACRO432030010EA 1500 SEK
ACRO432030010 ACRO432030050
trans-β-Styrylboronic acid pinacol ester 97%
trans-beta-Styrylboronic acid pinacol ester
Formel: C₁₄H₁₉BO₂
Kokpunkt: 89…93 °C (,1 mmHg)
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD03453666
CAS nummer: 83947-56-2
Order Now
Specifikation testresultat
Appearance Colorless to light yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥94.0 %