1-Boc-isonipecotic acid ethyl ester 97%
Leverantör: Thermo Scientific
Varning
Synonyms: Ethyl N-BOC-4-piperidinecarboxylate, Ethyl N-Boc-piperidine-4-carboxylate
431500050 431500250
ACRO431500050EA 1625 SEK
ACRO431500050 ACRO431500250
1-Boc-isonipecotic acid ethyl ester 97%
1-Boc-isonipecotic acid ethyl ester
Formel: C₁₃H₂₃NO₄
MW: 257,33 g/mol
Kokpunkt: 120…135 °C (,5 mmHg)
Densitet: 1,046 g/cm³
Flampunkt: 110 °C
Storage Temperature: Omgivande
MDL Number: MFCD01763998
CAS nummer: 142851-03-4
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %