5-Bromo-1-methyl-1H-indole-3-carboxylic acid 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 1H-Indole-3-carboxylic acid, 5-bromo-1-methyl-, 5-bromo-1-methyl-1H-indole-3-carboxylicacid, 5-Bromo-1-methylindole-3-carboxylic acid
431170010 431170050
ACRO431170010EA 3550 SEK
ACRO431170010 ACRO431170050
5-Bromo-1-methyl-1H-indole-3-carboxylic acid 97%
5-Bromo-1-methyl-1H-indole-3-carboxylic acid
Formel: C₁₀H₈BrNO₂
MW: 254,083 g/mol
Smältpunkt: 255,4…255.7 °C
Storage Temperature: Omgivande
MDL Number: MFCD14706162
CAS nummer: 400071-95-6
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥99.0 %
Stabilizer 150 to 400 ppm BHT